missing translation for 'onlineSavingsMsg'
Learn More

Cesium nitrate, Ionization Buffer Solution, Specpure™, Thermo Scientific Chemicals

Product Code. 15426885
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1% Cs (as Cs2CO3) in 5% HNO3
CAS 7789-18-6
Quantity 100 mL
Molecular Formula CsNO3
MDL Number MFCD00010963
UN Number UN3264
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[Cs+]
IUPAC Name cesium;nitrate
Molecular Weight (g/mol) 194.909
PubChem CID 62674
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Cesium nitrate Ionization Buffer Solution
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'