missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium solution for ICP 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
64.00 € - 144.00 €
Specifications
Concentration | 1±0.02g/L |
---|---|
Type | Mass Spectrometry Standard |
Analyte or Component Names | Cs |
Solvent or Matrix | H2O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12993384
|
Fisher Chemical
|
100 mL | Bottle |
64.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12903394
|
Fisher Chemical
S 014 250 |
250 mL | Bottle |
96.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12913394
|
Fisher Chemical
S 014 500 |
500 mL | Bottle |
144.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
1±0.02g/L | |
Mass Spectrometry Standard | |
Cs | |
H2O |
Safety and Handling
H225; H304; H315; H336; H410
P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501
Danger
missing translation for 'shelfLife' : 24 months (12 months after opening)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title