missing translation for 'onlineSavingsMsg'
Learn More

Cesium sulfate, 99.9% (metals basis), Thermo Scientific Chemicals

78.69 € - 219.09 €

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
PubChem CID 25137
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11312508
SDS View Documents
Thermo Scientific Alfa Aesar
014559.14
25 g
78.69 €
25g
Please to purchase this item. Need a web account? Register with us today!
11302508
SDS View Documents
Thermo Scientific Alfa Aesar
014559.22
100 g
219.09 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cesium sulfate is used to prepare dense aqueous solutions, which finds application in isopycnic centrifugation especially for DNA purification and recovery. It acts as a catalyst for sulfur dioxide oxidation with vanadium or vanadium pentoxide. Further, it acts as a metal oxide catalyst enhancer for heterogeneous processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10294-54-9
361.87
FLJPGEWQYJVDPF-UHFFFAOYSA-L
25137
Cs2O4S
MFCD00010959
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
[Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

1010°C
White
99.99% (Metal basis)
MFCD00010959
Soluble in water. Insoluble in ethanol and acetone.
[Cs+].[Cs+].[O-]S([O-])(=O)=O
361.87
361.87
White powder
Cesium sulfate
10294-54-9
Powder
Cs2O4S
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
FLJPGEWQYJVDPF-UHFFFAOYSA-L
dicaesium(1+) sulfate
25137
Hygroscopic
4.243
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H361f-H373

missing translation for 'einecsNumber' : 233-662-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.