missing translation for 'onlineSavingsMsg'
Learn More

Cesium sulfate, 99+%, pure

Product Code. p-222844 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
10g
250g
25g
This item is not returnable. View return policy

Product Code. 10722495

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
PubChem CID 25137
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1900.0°C
Melting Point 1005.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 10 g
Assay Percent Range 99% min
Linear Formula Cs2SO4
Merck Index 15,2015
Solubility Information Solubility in water: 1820g/L (25°C). Other solubilities: practically insoluble in alcohol, aceton, pyridine
IUPAC Name dicesium;sulfate
Formula Weight 361.86
Percent Purity ≥99%
Grade Pure
Packaging Glass bottle
Density 4.24
Chemical Name or Material Cesium sulfate, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.