missing translation for 'onlineSavingsMsg'
Learn More

Cesium sulfate, 99%, Thermo Scientific Chemicals

Product Code. 11402788
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.

Solubility
Soluble in water. (1820 g/L) at 20°C

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water, reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
PubChem CID 25137
IUPAC Name dicesium;sulfate
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

Melting Point 1,010°C
Quantity 25 g
Merck Index 14,2016
Solubility Information Soluble in water. (1820g/L) at 20°C
Formula Weight 361.87
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 4.24
Chemical Name or Material Cesium sulfate
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Cesium sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium sulfate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.