missing translation for 'onlineSavingsMsg'
Learn More

Cesium tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11353598
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cesium tungsten oxide is used for making transparent insulation coating that has the best infrared transparent barrier feature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13587-19-4
Molecular Formula Cs2O4W
MDL Number MFCD00049438
Synonym Cesium tungstate

Specifications

Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information It is soluble in water.
Formula Weight 513.66
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cesium tungsten oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'