Learn More
Tocris Bioscience™ Cetrorelix Acetate
Potent GnRH receptor antagonist
Brand:  Tocris Bioscience™ 3536/1
184.15 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18044102
  • 233.00 € / 1mg
Explore more special offers
Description
Description
Cetrorelix Acetate is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist (KD = 0.202 nM, IC50 = 1.21 nM). Suppresses production of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) from the pituitary gland, which inhibits ovulation. Exhibits antiproliferative effects and displays efficacy against hormone-sensitive cancers in vivo. Also exhibits anxiolytic and antidepressant activity in vivo.
Specifications
Specifications
Cetrorelix Acetate
Cetrorelix Acetate
Ac-D-2-Nal-D-(4-Cl-Phe)-D-(3-Pyridyl-Ala)-Ser-Tyr-D-Cit-Leu-Arg-Pro-D-Ala-NH2.CH3CO2H
1491.11