missing translation for 'onlineSavingsMsg'
Learn More

cGMP Polyclonal Antibody, Invitrogen™

Sheep Polyclonal Antibody

Brand:  Invitrogen PA110841

Product Code. 11505922

  • 499.00 € / 200µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

PA1-10841 detects cGMP. PA1-10841 has been successfully used in ELISA applications. The PA1-10841 immunogen is Succinylated cGMP, conjugated to BSA.

Guanosine cyclic 3',5'-(hydrogen phosphate) is a guanine nucleotide containing one phosphate group which is esterified to the sugar moiety in both the 3'- and 5'-positions. Cyclic gmp is a cellular regulatory agent and has been described as a second messenger. Its levels increase in response to a variety of hormones, including acetylcholine, insulin, and oxytocin and it has been found to activate specific protein kinases. [National Center for Biotechnology Information. PubChem Compound Database; CID=24316].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

cGMP
Polyclonal
Unconjugated
C10H12N5O7P; Cyclic GMP
Succinylated cGMP, conjugated to BSA.
RUO
Chemical
Antibody
IgG
ELISA
Conc. Not Determined
whole serum with 0.1% sodium azide
Sheep
200 μL
Primary
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers