missing translation for 'onlineSavingsMsg'
Learn More

CGP 52432, Tocris Bioscience™

Product Code. 30001215
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective GABAB antagonist

Potent, selective GABAB receptor antagonist (IC50 = 85 nM).

Chemical Identifiers

CAS 139667-74-6
Molecular Formula C15H24Cl2NO4P
Molecular Weight (g/mol) 384.234
InChI Key GJZVQXWEIYRHBE-UHFFFAOYSA-N
Synonym tocris-1246, biomol-nt_000239, 3-3,4-dichlorophenyl methyl amino propyl diethoxymethyl phosphinic acid, 3-3,4-dichlorphenyl methyl amino propyl diethoxymethyl phosphinic acid, 3-3,4-dichlorophenyl methylamino propyl-diethoxymethyl phosphinic acid, phosphinic acid, 3-3,4-dichlorophenyl methyl amino propyl diethoxymethyl
PubChem CID 132252
IUPAC Name 3-[(3,4-dichlorophenyl)methylamino]propyl-(diethoxymethyl)phosphinic acid
SMILES CCOC(OCC)P(=O)(CCCNCC1=CC(=C(C=C1)Cl)Cl)O

Specifications

Quantity 50 mg
Formula Weight 384.24
Percent Purity >99%
Chemical Name or Material CGP 52432
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.