missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CGP 71683 hydrochloride
Highly selective and potent non-peptide NPY Y5 receptor antagonist
304.00 € - 1180.00 €
Specifications
Purity | >99% |
---|---|
Product Type | CGP 71683 hydrochloride |
Molecular Weight (g/mol) | 512.07 |
Inhibitors | CGP 71683 hydrochloride |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18005412
|
Tocris Bioscience™
2199/50 |
50 mg |
1180.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18757227
|
Tocris Bioscience™
2199/10 |
10 mg |
304.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Extremely selective, non-peptide NPY Y5 receptor antagonist. Displays > 1000-fold selectivity over Y1, Y2 and Y4 receptors; IC50 values are 1.4, 2765, 7187 and 5637 nM at cloned rat Y5, Y1, Y2 and Y4 receptors respectively. Potently inhibits NPY-induced food intake following i.p. administration in diabetic, free-feeding and fasted rats.Specifications
>99% | |
CGP 71683 hydrochloride | |
512.07 | |
CGP 71683 hydrochloride |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts