Learn More
Tocris Bioscience™ CGS 35066
Endothelin-converting enzyme (ECE) inhibitor
Brand:  Tocris Bioscience™ 2512/10
242.58 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18035462
  • 267.00 € / 10mg
Explore more special offers
Description
Description
CGS 35066 is a potent endothelin-converting enzyme (ECE) inhibitor that displays > 100-fold selectivity over neutral endopeptidase 24.11 (IC50 values are 22 and 2300 nM respectively). Blocks the hypertensive effects induced by big ET-1 in vitro and reduces the magnitude of cerebral vasospasm following subarachnoid hemorrhage (SAH).
Specifications
Specifications
10 mg
CGS 35066
349.28
CGS 35066