missing translation for 'onlineSavingsMsg'
Learn More

R&L Enterprises™ Chain Clamps

Provides support for articles from 0 to 150mm in diameter. R&L Enterprises™ Chain Clamps have jaws that are tightened by the use of a plastic-headed tension screw.

Brand:  R&L Enterprises™ 9100

Product Code. 12936826

  • 61.80 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Shaft diameter of 10mm
  • Shaft and tensioner parts are nickel plated
  • Chain is coated in rubber
  • With cork-lined jaws
Specifications

Specifications

1 Ea.
Rubber Covered Chain
Cork Lined
Chain Clamp
For use with 0 to 150mm diameter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.