missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm™ Chamber Furnace LF 1400 Series with B500 Controller

Combined insulation of refractory bricks in the corners and low heat storage and quickly cooling fiber material. With a wide variety of optional equipment, these chamber furnaces can be optimally adapted to your processes.

12435.00 € - 20635.00 €

Specifications

Controller Type B500
Temperature (Metric) 1400°C
View More Specs

Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
17327939
View Documents
Nabertherm™
LF0242NKNE
15 L
12435.00 €
Each
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
 
17307949
View Documents
Nabertherm™
LF0642NKNE
60 L
16660.00 €
Each
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
 
17347949
View Documents
Nabertherm™
LF1242NKNE
120 L
20635.00 €
Each
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
 
17367939
View Documents
Nabertherm™
LF0342NKNE
30 L
14180.00 €
Each
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
 
Description

Description

  • Maximum Temperature: 1400°C
  • Dual shell housing with rear ventilation, provides for low shell temperatures
  • Five-sided heating for very good temperature uniformity
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Controller mounted on furnace door and removable for comfortable operation
  • Protection of bottom heating and flat stacking surface provided by embedded SiC plate in the floor
  • High-quality non-classified fiber insulation with corner bricks for shorter heating and cooling times
  • Door with brick-on-brick seal, hand fitted
  • Short heating times due to high installed power
  • Side vent with bypass connection for exhaust pipe
  • Self-supporting arch for high stability and greatest possible protection against dust
  • Quick lock on door
  • Freely adjustable air slide intake in furnace floor
  • Stand included
  • Defined application within the constraints of the operating instructions
Specifications

Specifications

B500
1400°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nabertherm™ Chamber Furnace LF 1400 Series with B500 Controller

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.