missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAPS, 98%, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- CHAPS (3-((3-cholamidopropyl) dimethylammonio)-1-propanesulfonate) is a zwitterionic non-denaturing detergent
- It is suitable for applications which require protection of native protein conformation
Applications
- CHAPS can be used for the solubilization and purification of membrane proteins
- It is also used in conjunction with nonionic detergents like Triton X-100
- It can be used as a non-denaturing zwitterionic detergent for the study of membrane chemistry
Specifications
Specifications
| Melting Point | 267.2°C to 271.4°C |
| Color | White |
| Elemental Analysis | 4.46 to 4.65 % N |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Quantity | 25 g |
| Merck Index | 15,2043 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohols like methanol and ethanol |
| Formula Weight | 614.9 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction