missing translation for 'onlineSavingsMsg'
Learn More

Charcoal, Technical, Decolourising Powder, ActIVated, Fisher Chemical™

50.40 € - 340.00 €

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
MDL Number 133992
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11478103
View Documents Promotion Details
Fisher Chemical
C/4040/53
500 g Plastic powder jar
50.40 €
500g
Please to purchase this item. Need a web account? Register with us today!
10000050
View Documents Promotion Details
Fisher Chemical
C/4040/60
1 kg Plastic powder jar
102.00 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10010050
View Documents Promotion Details
Fisher Chemical
C/4040/65
5 kg Plastic keg
340.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7440-44-0
C
133992

Specifications

7440-44-0
Black
133992
Powder
3652°C
C
12g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Charcoal, Activated

Signal Word
  • Warning

Hazard Category
  • Self-heating substance Category 2

Hazard Statement
  • H252-Self-heating in large quantities; may catch fire.

Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Product Certifications