Learn More
Eppendorf™ Charger Stand 2, for Eppendorf Xplorer™/Xplorer™ plus
GREENER_CHOICE
Saves bench space and conveniently stores and charges your pipette.
Brand:  Eppendorf™ 3116000031
228.54 EUR valid until 2024-12-06
Use promo code "21633" to get your promotional price.
Product Code. 15735223
  • 289.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Carries and charges one Eppendorf Xplorer™ or Xplorer™ plus pipette
  • Robust design that securely holds and protects the instrument
  • Large rubber feet protect stand from liquids spilled on workbench
  • Cord wrap functionality for unused power cord
  • Requires mains/power adapter (not included); power adapter is supplied with Eppendorf Xplorer™ or Xplorer™ plus Pipette
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Charger Stand 2
Pipette Holder and Charger
One Eppendorf Xplorer™ or Xplorer plus pipette
Storing and charging pipettes