Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Brand:  Thermo Scientific™ FCR700TSXLE
 View more versions of this product
Product Code. 15738537
  • 2030.10 € / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed ink chart recorder
Services
Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Inkless Chart Recorder, Field Installed
Chart Recorder
TSX700 ultra-low freezers
Recorder