missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Brand: Thermo Scientific™ FCRP500TSXLE
This item is not returnable.
View return policy
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed inkless chart recorderServices
Warranty varies by country, please contact us for details
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Ink Chart Recorder, Field Installed | |
Chart Recorder | |
TSX500 ultra-low freezers | |
Recorder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction