Learn More
Lovibond™ Checkit™ Comparator Single Parameter Test Kit
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
Brand:  Lovibond™ 147130
 View more versions of this product
Product Code. 10580853
  • 70.55 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
  • Compact and lightweight
  • Provides the basis for a comprehensive and easy-to-use analysis system
  • Comparator Discs contain a continuous color scale for exact color matching between standard and sample
Specifications
Specifications
Comparator apparatus, color comparison discs, cells, stirring rod, reagents, instructions
1 Pack
Checkit™ Comparator Single Parameter Test Kit
30
4 to 10pH