missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Checkit™ Comparator Single Parameter Test Kit

Product Code. p-8013690
Click to view available options
Range:
0 to 1mg/L
4 to 10pH
5.2 to 6.8mg/L
6 to 7.6mg/L
Analytes:
Ammonia
Copper
pH
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10570853

Brand: Lovibond™ 147120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.

  • Compact and lightweight
  • Provides the basis for a comprehensive and easy-to-use analysis system
  • Comparator Discs contain a continuous color scale for exact color matching between standard and sample

Specifications

Analytes pH
For Use With (Application) For Both Mobile and Static Water Analysis
Includes Comparator apparatus, color comparison discs, cells, stirring rod, reagents, instructions
Quantity 1 Each
Range 6 to 7.6mg/L
Type Checkit™ Comparator Single Parameter Test Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.