missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total

Accurate test kits for routine water analysis

55.90 € - 95.85 €

Specifications

Accuracy ±5%
For Use With (Application) Routine water analysis
No. of Tests 100
Type Chlorine Free and Total Test Kit
View More Specs

Products 9
Product Code Brand Range Includes Price Quantity & Availability  
Product Code Brand Range Includes Price Quantity & Availability  
10094454
View Documents
Lovibond™
147052
0 to 3.5mg/L Aqua comparator™ housing, aqua comparator™ disc, DPD free reagent, 5mL sample, DPD total reagent, 5mL sample, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
95.85 €
Each
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
 
11745458
View Documents
Lovibond™
147020
0 to 4mg/L Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10700255
View Documents
Lovibond™
147020
0 to 4mg/L Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
67.20 €
Each
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
 
11775458
View Documents
Lovibond™
147040
0.1 to 2mg/L Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10769674
View Documents
Lovibond™
147040
0.1 to 2mg/L Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
67.20 €
Each
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
 
10326622
View Documents
Lovibond™
147000
0.02 to 0.3mg/L Aqua comparator™ housing, aqua comparator™ disc D55, DPD No. 1 tablets, DPD No. 3 tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
75.85 €
Each
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
 
11715458
View Documents
Lovibond™
147000
0.02 to 0.3mg/L Aqua comparator™ housing, aqua comparator™ disc D55, DPD No. 1 tablets, DPD No. 3 tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11725458
View Documents
Lovibond™
147010
0 to 1mg/L Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10663713
View Documents
Lovibond™
147010
0 to 1mg/L Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
67.20 €
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
 
Description

Description

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Specifications

±5%
Routine water analysis
100
Chlorine Free and Total Test Kit
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.