missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medimark International™ Chemgene HLD4L Laboratory Surface Disinfectant
Click to view available options
Volume (Metric):
750 mL
1 L
5 L
Unit Size:
1 L
5 L
750mL
Description
For multipurpose surface disinfection and cleaning. Chemgene™ Lab Disinfectant Spray destroys bacteria, fungi, mycobacteria, viruses and spores.
Chemgene HLD4L disinfectant concentrate.
- Manufactured in accordance with Quality Management System ISO 9001
- Biodegradable under OECD conditions
- Denatures DNA/RNA
Specifications
Specifications
| Product Type | Lab Disinfectant Spray |
| Appearance | Clear |
| Dilution | 1 in 100, General conditions; or 1 in 10 for Bio-hazard contamination |
| For Use With (Application) | For multipurpose Surface disinfection and cleaning |
| Volume (Metric) | 750 mL |
Safety and Handling
Product Identifier
- CHEMGENE HLD4L 1:20 RTU TRIGGER SPRAY
- Danger||
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
- P102-Keep out of reach of children.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P315-Get immediate medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction