missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX ChemiBrite™ FSH Glycoprotein Hormone Receptor Stable Cell Line

Product Code. 17325329
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17325329

Brand: DiscoverX HTS178L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ChemiBrite™ FSH Glycoprotein Hormone Receptor Stable Cell Line measures FSHR (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.

The ChemiBrite FSH Glycoprotein Hormone Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. follicle stimulating hormone) based activation of FSHR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses FSHR alongside exogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of FSHR , the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Calcium Flux
Cell Line 293H
For Use With (Application) Cell Lines
Primary Agonist FSH, Human
Quantity 2 Vials
Target Entry FSHR
Accession Number NM_000145
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.