Learn More
Brady™ Chemical-resistant Cryogenic Polypropylene Laboratory Labels for M6/M7 Printers
Polypropylene material is ideal for general purpose barcode applications and withstands liquid nitrogen, autoclave and other harsh environments.
Brand:  Brady™ 174078
 View more versions of this product
Product Code. 17858404
  • 84.90 € / Pack of 100
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Matte white polypropylene with acrylic adhesive is thermal transfer printable
  • Ideal for laboratory identification such as vials, centrifuge tubes and test tubes
  • Service temperature of -80°C to 70°C (-112°F to 158°F)
  • Made of B-7425 polypropylene material
Specifications
Specifications
Autoclavable
M6-121-7425
None
Matte
Polypropylene
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
100 Labels
-112°F to +158°F
0.004 in.
1.5 x 1.9 in.
1.9 in.
38.1 mm
ASTM D 1000, ASTM D 2979, RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Permanent Acrylic
White
Die-cut
Single-side Printable
Labels
Equipment Labeling, General Identification, Laboratory Labeling, Temperature Indicating Labeling
No
Rectangle
-80°C to +70°C
0.102 mm
1.5 in.
48.26 mm