missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Chemical Spill Kit
Ensures that everything is available, and ready should a spill occur. Cleaning up hazardous chemical spills quickly in an emergency situation promotes on-site safety.
509.00 € - 1275.00 €
Specifications
Type | Chemical Spill Response Kit |
---|---|
Format | Kit |
For Use With (Application) | Rapid Response to Chemical Spills |
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
17139350
|
3M™
7100090374 |
10 x P110 Sheets, 1 x P300 Pillow, 1 disposal bag with ties |
509.00 €
5 sets |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
17129350
|
3M™
7100090074 |
(100) P110 Sheets, (12) P300 pillows, (8) P200 Mini-booms, 5 disposal bags with ties |
1275.00 €
1 set |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
Description
- Ideal for rapid response to chemical spills: fast and effective containment and clean-up
- Cleaning up hazardous chemical spills, drips, and leaks
- Suitable for use with most chemicals
- Promotes on-site safety
- Easy to dispose of
Specifications
Chemical Spill Response Kit | |
Kit | |
Rapid Response to Chemical Spills |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Chemical Spill Kit