Learn More
Thermo Scientific™ Cheminert Model C2 Microbore Injector
Can be used as an injector or switching valve
65.65 €
Product Code | Brand | Volume (Metric) Sample | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) Sample | Price | Quantity & Availability | |||||
11523970
|
Thermo Scientific™
CZSL10PK |
10 μL | N/A | ||||||
Description
- 1/16 in. fittings
- 0.010 in. ports
- Available in 6-port or 10-port configurations
- Available with manual or microelectric actuator