Learn More
Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors
Product Code. p-4522328 Shop All Thermo Scientific Products
Click to view available options
Stator Material:
N60 Stainless
Actuation:
Microelectric
No. of Ports:
10 Ports
Unit Size:
Each
Product Code. 10289274
Brand: Thermo Scientific™ C21000EP
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5μL sample loop.
  • 1/16″ fittings
  • 0.010″ ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator
TRUSTED_SUSTAINABILITY
Specifications
Actuation Microelectric
Volume (Metric) Sample 5 μL Loop
For Use With (Equipment) HPLC
Type Microbore Injector/Switching Valve
No. of Ports 10 Ports
Stator Material N60 Stainless
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.