Learn More
VACUUBRAND™ Chemistry Pumping Unit, PC 620 Select
GREENER_CHOICE
For simultaneous operation of two electronically controlled vacuum applications with one single pump. Similarly, Models 511 and 611 Select have one port that is manually controlled. A VACUU·SELECT Vacuum Controller for a simplified work flow.
Brand:  VACUUBRAND™ 20737352
 View more versions of this product
Product Code. 16231792
  • 10870.00 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • VACUU·SELECT Vacuum Controller with graphical user interface simplifies work in the laboratory
  • Predefined applications save time and give reproducible results
  • Easy-to-use application editor for the creation of unique process sequences
  • Powerful vacuum pump with excellent chemical and condensate compatibility
  • Chemical resistant solenoid valves to maintain process control
  • Pressure is maintained at the level where the first boiling point is detected by the controller
  • Glass inlet separator with a robust protective coating prevents particles and liquid droplets from entering the pump, preventing damage and extending pump life
  • Exhaust vapor condenser allows for near complete recovery of solvents when used with a chilled water line or recirculating chiller
  • Use manual set-point-control for simple processes
  • Meets high vacuum requirements for most high boiling solvents
  • Touch screen display can be operated even when wearing safety gloves
  • Choose from 14 languages
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.5 Mbar (Without Gas Ballast), 3 Mbar (With Gas Ballast)
24.6 kg
1500 to 1800 min-1
435 x 243 x 457 mm
PC 620 Select
UK
21.2 kg
10°C
50-60 Hz
IP40
Rotary Evaporator/Parallel Evaporator, Vacuum Concentrators, Drying Chambers
1.1 bar
VACUU·SELECT
Ready-to-Use Pumping Unit (Completely Mounted with Manual)
CE-certification; II 3/-G IIC T3 x Internal Atm. Only
PTFE (Partly Carbon Reinforced), ETFE/ECTFE (Partly Carbon Fiber Reinforced), FFKM
45 dBA
0.25 kW
40°C
3.4 m3/hr.
Diaphragm Pump
4
3
230 V