Learn More
Thermo Scientific Chemicals Chemotactic Peptide
Brand:  Thermo Scientific Chemicals J65184.MB
Product Code. 15496769
  • 121.61 € / 25mg
Explore more special offers
Description
Description
Induces leucocyte chemotaxis, and activates macrophages
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HPLC
-30°C to -10°C
Induces leucocyte chemotaxis, and activates macrophages
Chemotactic Peptide
RUO - research use only
fMLP
Lyophilized Powder
437.55
25 mg
N-Formyl-Met-Leu-Phe