Learn More
Ibidi Chemotaxis Sticky-Slide
Chemotaxis Sticky-Slide is a ready-to-use bottomless slide for chemotaxis assays with fast or slow migrating cells with a self-adhesive underside to which own substrates can be mounted.
Brand:  Ibidi 80328
Encompass
Product Code. 17216173
  • POA / Pack of 10
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • A ready-to-use microscopy slide for chemotaxis assays with fast or slow migrating cells
  • suitable for 2D or 3D cell culture
  • stable chemotactic gradients for long-term experiments
  • bottomless with self-adhesive underside
  • sterilized
Specifications
Specifications
Sterile
1
10
Microscope Slide
Chemotaxis Sticky Slide
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ibidi Chemotaxis Sticky-Slide