missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ ChemTek™ 38-520 Series Butyl Polymer Immersion Gloves
Provides the best resistance to the most aggressive chemicals without compromising dexterity or comfort. ChemTekTM gloves are surprisingly flexible and provide excellent dexterity.
90.00 €
Specifications
Back Material | Butyl |
---|---|
Cuff Material | Butyl |
Coating Material | Butyl Polymer |
Laboratory/Cleanroom Use | Yes |
Latex Free | Yes |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12629711
|
Ansell™
38520100 |
10 |
90.00 €
1 pair |
In Stock
Log in to see stock. |
|||||
12649711
|
Ansell™
38520070 |
7 |
90.00 €
1 pair |
In Stock
Log in to see stock. |
|||||
12639711
|
Ansell™
38520110 |
11 |
90.00 €
1 pair |
In Stock
Log in to see stock. |
|||||
12659711
|
Ansell™
38520080 |
8 |
90.00 €
1 pair |
In Stock
Log in to see stock. |
|||||
12669711
|
Ansell™
38520090 |
9 |
90.00 €
1 pair |
In Stock
Log in to see stock. |
|||||
Description
- Smooth finish in black
- Easy donning and good grip
- Maximum protection from hazardous chemicals
- Surprisingly flexible and provide excellent dexterity
- Designed for comfort with a natural, curved ergonomic shape and soft feel
- Absence of latex proteins means there's no risk of latex allergies
- Ideal for the chemical industry, oil and petrol refining, printing industry, automotive industry, mining and aerospace
Specifications
Butyl | |
Butyl Polymer | |
Yes | |
38-520 | |
Black | |
Black | |
Individually wrapped bags with insert | |
Highly chemical resistance |
Butyl | |
Yes | |
Immersion Gloves | |
20 mil | |
Beaded/Rolled | |
Yes | |
1 Pair | |
38-520 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title