Learn More
Fisherbrand™ Chest ULT Freezer Racks
Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of chest ULT freezers.
Brand:  Fisherbrand™ CF-11-2
 View more versions of this product
Product Code. 15789529
  • 85.60 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Holds standard cryoboxes
  • Made of high quality stainless steel
  • Locking rod included
Specifications
Specifications
Vertical Rack
Stainless Steel
Chest ULT Freezer
2 in. Boxes (11)
Standard 2 in. Cryoboxes
615 x 143 x 140mm