Learn More
Invitrogen™ Chicken anti-Bovine IgG (H+L) Secondary Antibody, HRP
Chicken Polyclonal Secondary Antibody
Brand: Invitrogen™ SA172015
126.67 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
SA1-72015 detects Bovine IgG in bovine samples. SA1-72015 has been successfully used in ELISA and Western blot procedures. The SA1-72015 immunogen is from purified bovine IgG isolated from normal pooled bovine serum.
Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Bovine IgG (H+L) | |
Polyclonal | |
HRP | |
Chicken | |
purified | |
RUO | |
Bovine | |
Secondary Antibody | |
IgY |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal | |
Purified bovine IgG isolated from normal pooled bovine serum. | |
500 μg | |
Secondary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.