missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chicken anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, AP
Chicken Polyclonal Secondary Antibody
Brand: Invitrogen™ A15996
This item is not returnable.
View return policy
Description
Chicken anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody for Western Blot, IHC, ELISA
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Chicken | |
Affinity chromatography | |
RUO | |
Rabbit | |
Secondary Antibody | |
IgY |
Western Blot, ELISA, Immunohistochemistry | |
2.5 mg/mL | |
secondary antibody with 1% BSA, 5mM MgCl2, 30mM triethanolamine, 0.1mM ZnCl2 and 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains. | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Chicken anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, AP > 500 μg; Alkaline Phosphatase (AP)
Spot an opportunity for improvement?Share a Content Correction