Learn More
Invitrogen™ Chicken anti-Rat IgG (H+L) Secondary Antibody, Biotin
Chicken Polyclonal Secondary Antibody
Brand:  Invitrogen™ PA130361
Product Code. 11541363
  • 489.00 € / 1mg
Explore more special offers
Description
Description
PA1-30361 detects Rat IgG from rat samples.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rat IgG (H+L)
Polyclonal
Biotin
Chicken
IgG fraction
RUO
Rat
Secondary Antibody
IgY
Immunohistochemistry, ELISA, Western Blot
1 mg/mL
0.02M potassium phosphate with 10mg/mL BSA, 0.15M NaCl and 0.01% sodium azide; pH 7.2
Rat IgG whole molecule.
1 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Chicken anti-Rat IgG (H+L) Secondary Antibody, Biotin > 1 mg; Biotin