Learn More
Novus Biologicals™ Chicken Universal Protein Whole Tissue Lysate (Adult Whole Normal)
Brand:  Novus Biologicals™ NBL1-26503
Product Code. 18197489
  • 412.00 € / 2 units
Explore more special offers
Description
Description
The Chicken Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from chicken.
Specifications
Specifications
Universal
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Chicken
Chicken Universal Protein Whole Tissue Lysate (Adult Normal)
Adult
2 Units
Universal Protein Lysate: Chicken Normal Tissues
Normal
Chicken
-80°C. Avoid freeze-thaw cycles.
Native