Learn More
Sherwood Scientific™ M926S Chloride Analyzer
Analyze chloride content via reliable colorimetric titration methodology. Sherwood Scientific™ M926S Chloride Analyzer is an ergonomically designed and microprocessor-based instrument.
Brand:  Sherwood Scientific™ 92600109
Product Code. 12876813
  • 7500.00 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Colorimetric titration method where silver ions are generated in-situ and react with chloride ions.
  • Accurate, proven method for chloride and salt concentration.
  • Wide range of applications including those in clinical (Model 926S) and food industries.
  • Very easy to use.
  • Equipped with RS232 serial and USB outputs which enable them to be linked to a suitable printer or to be interfaced to a PC hosting Active Salt software.
  • Active Salt package can collect data directly from a balance with RS232 output together with the chloride analyser results.
  • Software may be configured by the operator to suit their particular laboratory requirements.
  • DataGate LIMS and auto reading of balance and barcode.
  • No transcription errors
  • No time wasted doing manual calculations.
  • No need to achieve specific sample and diluent weights.
  • Automatic data storage and analysis report generation.
  • End point precisely detected.
  • No reagent needing recalibration.
  • Economical per test.
Specifications
Specifications
20 μL to 100μL
Printer cable, acid buffer, standard, 3 anodes, set of electrodes, 2 marked beakers, electrode cleaning polish, stirrer and manual
100 to 240 V