Learn More
Palintest™ Chlorine DPD 1 Reagent
Robust, reliable, replacement ChloroSense sensors for use with ChloroSense meter, 99359-10.
Brand: Palintest™ PM011
This item is not returnable.
View return policy
Description
- The simplest, portable method for accurate measurements of free and total chlorine - with USEPA approval
- X500 replacement electrode
- 500 replacement sensors for free and total chlorine
- Sensors individually wrapped in desiccant foil
Specifications
50 Pack |