missing translation for 'onlineSavingsMsg'
Learn More

Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh, Thermo Scientific Chemicals

Product Code. 10266600
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12092-47-6
Molecular Formula C16H24Cl2Rh2
Molecular Weight (g/mol) 493.08
MDL Number MFCD00012415
InChI Key QSUDXYGZLAJAQU-MIXQCLKLSA-L
Synonym chloro 1,5-cyclooctadiene rhodium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, 1,5-cyclooctadiene rhodium chloride dimer, ccris 5036, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer
PubChem CID 6436379
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 256.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: 0.34g/L
Formula Weight 493.08
Percent Purity ≥40.8% (RH)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Chloro(1, 5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'