missing translation for 'onlineSavingsMsg'
Learn More

Chloro(methyl)diphenylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11386816
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chloro(methyl)diphenylsilane is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloro(methyl)diphenylsilane is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong bases. Incompatible with moisture and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-79-6
Molecular Formula C13H13ClSi
Molecular Weight (g/mol) 232.78
MDL Number MFCD00000498
InChI Key OJZNZOXALZKPEA-UHFFFAOYSA-N
Synonym chloro methyl diphenylsilane, methyldiphenylchlorosilane, chloromethyldiphenylsilane, silane, chloromethyldiphenyl, chlorodiphenylmethylsilane, diphenylmethylchlorosilane, methyldiphenylsilyl chloride, dpmscl, chloro-methyl-diphenyl-silane, benzene, 1,1'-chloromethylsilylene bis
PubChem CID 67349
IUPAC Name chloro-methyl-diphenylsilane
SMILES C[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 294°C to 295°C
Quantity 50 g
UN Number UN2987
Beilstein 2937445
Flash Point 141°C (285°F)
Solubility Information Reacts with water.
Formula Weight 232.79
Percent Purity 97%
Odor Irritating
Refractive Index 1.574
Sensitivity Moisture sensitive
Density 1.104
Chemical Name or Material Chloro(methyl)diphenylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'