Learn More
Chloroacetonitrile, 98+%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 107-14-2 |
---|---|
Molecular Formula | C2H2ClN |
Molecular Weight (g/mol) | 75.495 |
MDL Number | MFCD00001885 |
InChI Key | RENMDAKOXSCIGH-UHFFFAOYSA-N |
Synonym | chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro |
PubChem CID | 7856 |
ChEBI | CHEBI:82416 |
IUPAC Name | 2-chloroacetonitrile |
SMILES | C(C#N)Cl |
Description
Chloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen's condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen′s condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and acids.
Chemical Identifiers
107-14-2 | |
75.495 | |
RENMDAKOXSCIGH-UHFFFAOYSA-N | |
7856 | |
2-chloroacetonitrile |
C2H2ClN | |
MFCD00001885 | |
chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro | |
CHEBI:82416 | |
C(C#N)Cl |
Specifications
107-14-2 | |
124°C to 126°C | |
≥98% | |
1.423 | |
506028 | |
Immiscible with water. | |
C(C#N)Cl | |
75.495 | |
CHEBI:82416 | |
Liquid |
1.195 | |
56°C (132°F) | |
C2H2ClN | |
MFCD00001885 | |
chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro | |
RENMDAKOXSCIGH-UHFFFAOYSA-N | |
2-chloroacetonitrile | |
7856 | |
75.5 | |
Chloroacetonitrile |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P363-P370+P378q-P501c
H226-H301+H331-H310-H319
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: I
missing translation for 'einecsNumber' : 203-467-0
missing translation for 'rtecsNumber' : AL8225000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.