missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2500 mL
Description
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.
Solubility
Miscible with alcohol, chloroform, diethyl ether and benzene. Slightly immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.
Solubility
Miscible with alcohol, chloroform, diethyl ether and benzene. Slightly immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
Melting Point | -45°C |
Density | 1.106 |
Boiling Point | 132°C |
Quantity | 500 mL |
Odor | Aromatic |
Refractive Index | 1.524 |
UN Number | UN1134 |
Beilstein | 605632 |
Merck Index | 14,2121 |
Solubility Information | Miscible with alcohol,chloroform,diethyl ether and benzene. Slightly immiscible with water. |
Show More |
Safety and Handling
Product Identifier
- Chlorobenzene
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Chlorobenzene
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'