missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic-coated Glass Bottle
Specifications
Specifications
Melting Point | -45°C |
Boiling Point | 131°C |
Quantity | 2.5 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 12mbar at 20°C |
Viscosity | 0.8 mPaS at 20°C |
Formula Weight | 112.56g/mol |
Safety and Handling
Product Identifier
- Chlorobenzene
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyA§ thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/A§/if you feel unwell.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction