Learn More
Chlorobis(cyclooctene)iridium(I) dimer, Ir nominally 42.9%, Thermo Scientific Chemicals
Description
Chlorobis(cyclooctene)iridium(I) dime is used as catalyst for Isomerization-hydroboration reactions with nido-carboranyldiphosphine as stabilizing ligand, Hydrogen peroxide oxidation of hydroxamic acids and their subsequent hetero Diels-Alder cycloaddition reactions, Immobilization of organic functional groups onto solid supports through vinylsilane coupling reactions, Alkylation reactions, Guerbet reaction, Allylic amination reactions in a DNA-diene-iridium(I) hybrid system, Asymmetric hydroamination reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 5 g |
Solubility Information | Soluble in organic solvents. |
Formula Weight | 896 |
Concentration or Composition (by Analyte or Components) | Ir nominally 42.9% |
Chemical Name or Material | Chlorobis (cyclooctene)iridium (I) Dimer |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.