missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(ethylene)rhodium(I) dimer, 99%, Thermo Scientific Chemicals

Product Code. 15411537
Click to view available options
Quantity:
1 g

 

Chlorobis(ethylene)rhodium(I) dimer is used as a precursor to rhodium(I) norbornadiene catalyst, which is used in an asymmetric 1,4-addition of arylboronic acids to cyclic enones. Further, it serves as an important organic intermediate. It catalyzes the dimerization of ethylene to 1-butene. It is also used in the preparation of homogeneous catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 12081-16-2
Molecular Formula C8H16Cl2Rh2
MDL Number MFCD00013206
Quantity 1 g
Sensitivity Air Sensitive
Synonym chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i
Solubility Information Sparingly soluble in ethanol,acetone,dioxane,chloroform and dichloromethane. Insoluble in water.
InChI Key OOQJCPOXJFCGCR-UHFFFAOYSA-L
SMILES Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C
IUPAC Name bis(chlororhodium); tetrakis(ethene)
Molecular Weight (g/mol) 388.93
PubChem CID 45357812
Formula Weight 388.93
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Chlorobis(ethylene)rhodium(I) dimer
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'