missing translation for 'onlineSavingsMsg'
Learn More

chlorodiisopropylphosphine, 96%

Product Code. 10605731 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10605731

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40244-90-4 | C6H14ClP | 152.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40244-90-4
Molecular Formula C6H14ClP
Molecular Weight (g/mol) 152.61
MDL Number MFCD00015027
InChI Key JZPDBTOWHLZQFC-UHFFFAOYSA-N
Synonym chlorodiisopropylphosphine, diisopropylchlorophosphine, di-i-propylchlorophosphine, phosphinous chloride, bis 1-methylethyl, ipr2pcl, pubchem6476, chlorodiisopropylphosphane, chloro diisopropylphosphine, acmc-209jc6, diisopropylphosphinyl chloride
PubChem CID 538967
IUPAC Name chloro-di(propan-2-yl)phosphane
SMILES CC(C)P(C(C)C)Cl

Specifications

Color Yellow
Density 0.9500g/mL
Boiling Point 69.0°C (33.0 mmHg)
Flash Point 3°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula [(CH3)2CH]2PCl
Refractive Index 1.4740 to 1.4760
Quantity 5 g
Specific Gravity 0.95
Formula Weight 152.61
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Chlorodiisopropylphosphine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chlorodiisopropylphosphine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.