missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethyl-3,3,3-trifluoropropylsilane, 96%, Thermo Scientific™

Product Code. 11379305
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1481-41-0
Molecular Formula C5H10ClF3Si
Molecular Weight (g/mol) 190.665
MDL Number MFCD00053211
InChI Key KBAZUXSLKGQRJF-UHFFFAOYSA-N
Synonym trifluoropropyldimethylchlorosilane, chlorodimethyl-3,3,3-trifluoropropylsilane, 3,3,3-trifluoropropyl dimethylchlorosilane, chlorodimethyl 3,3,3-trifluoropropyl silane, silane, chlorodimethyl 3,3,3-trifluoropropyl, dimethyl 3,3,3-trifluoropropyl chlorosilane, chloro-dimethyl-3,3,3-trifluoropropyl silane, acmc-1c1aj, chloro dimethyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropylchlorodimethylsilane
PubChem CID 73866
IUPAC Name chloro-dimethyl-(3,3,3-trifluoropropyl)silane
SMILES C[Si](C)(CCC(F)(F)F)Cl

Specifications

Boiling Point 118°C to 120°C
Quantity 25 g
UN Number UN2985
Beilstein 1741726
Flash Point 20°C (68°F)
Formula Weight 190.67
Percent Purity 96%
Refractive Index 1.375
Sensitivity Moisture sensitive
Density 1.105
Chemical Name or Material Chlorodimethyl-3,3,3-trifluoropropylsilane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado