missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethyl-n-octadecylsilane, 95%, Thermo Scientific Chemicals

Product Code. 11384568
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Preparation of hydrophobic surfaces, chain terminator

Chlorodimethyl-n-octadecylsilane is used for preparation of hydrophobic surfaces and chain terminator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18643-08-8
Molecular Formula C20H43ClSi
Molecular Weight (g/mol) 347.10
MDL Number MFCD00000504
InChI Key GZGREZWGCWVAEE-UHFFFAOYSA-N
Synonym chlorodimethyloctadecylsilane, dimethyloctadecylchlorosilane, chlorodimethyl octadecyl silane, octadecyldimethylchlorosilane, silane, chlorodimethyloctadecyl, n-octadecyldimethylchlorosilane, chloro dimethyl octadecylsilane, chlorodimethyl-n-octadecylsilane, dimethyl-n-octadecylchlorosilane, stearyldimethylchlorosilane
PubChem CID 87738
IUPAC Name chloro-dimethyl-octadecylsilane
SMILES CCCCCCCCCCCCCCCCCC[Si](C)(C)Cl

Specifications

Boiling Point 184°C to 186°C (0.2mmHg)
Melting Point 28°C to 32°C
Color White
Physical Form Solid
Quantity 5 g
UN Number UN3261
Beilstein 2246012
Flash Point 200°C (392°F)
Solubility Information Reacts with water.
Formula Weight 347.1
Percent Purity 95%
Odor Acrid
Refractive Index 1.453
Sensitivity Moisture sensitive
Density 0.850 g/mL
Chemical Name or Material Chlorodimethyl-n-octadecylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.