missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethyl-n-octadecylsilane, 95%, Thermo Scientific Chemicals

Product Code. 11394568
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Preparation of hydrophobic surfaces, chain terminator

Chlorodimethyl-n-octadecylsilane is used for preparation of hydrophobic surfaces and chain terminator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18643-08-8
Molecular Formula C20H43ClSi
Molecular Weight (g/mol) 347.10
MDL Number MFCD00000504
InChI Key GZGREZWGCWVAEE-UHFFFAOYSA-N
Synonym chlorodimethyloctadecylsilane, dimethyloctadecylchlorosilane, chlorodimethyl octadecyl silane, octadecyldimethylchlorosilane, silane, chlorodimethyloctadecyl, n-octadecyldimethylchlorosilane, chloro dimethyl octadecylsilane, chlorodimethyl-n-octadecylsilane, dimethyl-n-octadecylchlorosilane, stearyldimethylchlorosilane
PubChem CID 87738
IUPAC Name chloro-dimethyl-octadecylsilane
SMILES CCCCCCCCCCCCCCCCCC[Si](C)(C)Cl

Specifications

Boiling Point 184°C to 186°C (0.2mmHg)
Melting Point 28°C to 32°C
Color White
Physical Form Solid
Quantity 100 g
UN Number UN3261
Beilstein 2246012
Flash Point 200°C (392°F)
Solubility Information Reacts with water.
Formula Weight 347.1
Percent Purity 95%
Odor Acrid
Refractive Index 1.453
Sensitivity Moisture sensitive
Density 0.850 g/mL
Chemical Name or Material Chlorodimethyl-n-octadecylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'