Learn More
Chlorodiphenylsilane, tech. 90%, Thermo Scientific Chemicals
77.69 € - 253.26 €
Chemical Identifiers
CAS | 1631-83-0 |
---|---|
Molecular Formula | C12H10ClSi |
Molecular Weight (g/mol) | 217.747 |
MDL Number | MFCD00045144 |
InChI Key | YCITZMJNBYYMJO-UHFFFAOYSA-N |
Synonym | chlorodiphenylsilane, diphenylchlorosilane, chloro diphenyl silane, silane, chlorodiphenyl, benzene, 1,1'-chlorosilylene bis, c12h11clsi, chlorodiphenylsilyl, diphenylsilyl chloride, chloro diphenyl silicon |
PubChem CID | 6327323 |
IUPAC Name | chloro(diphenyl)silicon |
SMILES | C1=CC=C(C=C1)[Si](C2=CC=CC=C2)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392226
|
Thermo Scientific Alfa Aesar
L03602.06 |
5 g |
77.69 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302236
|
Thermo Scientific Alfa Aesar
L03602.14 |
25 g |
253.26 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from bases, water. Incompatible with oxidizing agents and water.
Chemical Identifiers
1631-83-0 | |
217.747 | |
YCITZMJNBYYMJO-UHFFFAOYSA-N | |
6327323 | |
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)Cl |
C12H10ClSi | |
MFCD00045144 | |
chlorodiphenylsilane, diphenylchlorosilane, chloro diphenyl silane, silane, chlorodiphenyl, benzene, 1,1'-chlorosilylene bis, c12h11clsi, chlorodiphenylsilyl, diphenylsilyl chloride, chloro diphenyl silicon | |
chloro(diphenyl)silicon |
Specifications
280°C | |
C12H10ClSi | |
UN2987 | |
105°C (221°F) | |
Reacts with water. | |
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)Cl | |
217.747 | |
218.76 | |
Technical | |
Moisture sensitive | |
Chlorodiphenylsilane |
1631-83-0 | |
MFCD00045144 | |
2937164 | |
chlorodiphenylsilane, diphenylchlorosilane, chloro diphenyl silane, silane, chlorodiphenyl, benzene, 1,1'-chlorosilylene bis, c12h11clsi, chlorodiphenylsilyl, diphenylsilyl chloride, chloro diphenyl silicon | |
YCITZMJNBYYMJO-UHFFFAOYSA-N | |
chloro(diphenyl)silicon | |
6327323 | |
90% | |
1.579 | |
1.123 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 216-634-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Chlorodiphenylsilane
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Chlorodiphenylsilane
RUO – Research Use Only